ENA048303511 Enamine

2-chloro-3-(chloromethyl)-7-(methylsulfanyl)quinoline

  • MDL number MFCD06368065
  • Cas Number : 794582-34-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 125-126
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11507

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11507
Availability details for
Quantity
  • Availability
  • Add to Cart