ENA048303513 Enamine

2,3,5,6-tetramethylbenzenesulfinic acid

  • MDL number MFCD06368062
  • Cas Number : 957029-02-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11509

Safety Information

msds https://www.enaminestore.com/catalog/EN300-11509
Availability details for
Quantity
  • Availability
  • Add to Cart