ENA048303515 Enamine

2-methyl-3-(piperazin-1-ylmethyl)imidazo[1,2-a]pyridine

  • MDL number MFCD06368069
  • Cas Number : 794582-36-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 163...165
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11513

Safety Information

msds https://www.enaminestore.com/catalog/EN300-11513
Availability details for
Quantity
  • Availability
  • Add to Cart