ENA048303516 Enamine

6-bromo-3-(3-ethoxypropyl)-2-mercaptoquinazolin-4(3H)-one

  • MDL number MFCD06368754
  • Cas Number : 795290-91-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11515

Safety Information

msds https://www.enaminestore.com/catalog/EN300-11515
Availability details for
Quantity
  • Availability
  • Add to Cart