ENA048303523 Enamine

3-(N-benzyl4-chloro-2-fluorobenzenesulfonamido)propanoic acid

  • MDL number MFCD06368273
  • Cas Number : 794584-40-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11523

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11523
Availability details for
Quantity
  • Availability
  • Add to Cart