ENA048303526 Enamine

(2E)-3-(5-methoxy-2-methyl-2,3-dihydro-1-benzofuran-6-yl)prop-2-enoic acid

  • MDL number MFCD06368744
  • Cas Number : 795287-64-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11527

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11527
Availability details for
Quantity
  • Availability
  • Add to Cart