ENA048303534 Enamine

ethyl 4-acetyl-2-(2-chloroacetamido)-5-methylfuran-3-carboxylate

  • MDL number MFCD06368759
  • Cas Number : 795290-93-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11540

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11540
Availability details for
Quantity
  • Availability
  • Add to Cart