ENA048303548 Enamine

ethyl 4-amino-2-(chloromethyl)-6-methylfuro[2,3-d]pyrimidine-5-carboxylate

  • MDL number MFCD06370769
  • Molecular Weight : 269.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 188-189
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-11564

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11564
Availability details for
Quantity
  • Availability
  • Add to Cart