ENA048303549 Enamine

2-{[4-(2,4-dimethylbenzenesulfonamido)phenyl]formamido}acetic acid

  • MDL number MFCD06371692
  • Cas Number : 796083-83-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-11565

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11565
Availability details for
Quantity
  • Availability
  • Add to Cart