ENA048303551 Enamine

2-(5-acetamido-2-methoxybenzenesulfonamido)-3-phenylpropanoic acid

  • MDL number MFCD06655244
  • Cas Number : 87670-83-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-11567

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11567
Availability details for
Quantity
  • Availability
  • Add to Cart