ENA048303557 Enamine

[(5-chlorothiophen-2-yl)methyl](ethyl)amine

  • MDL number MFCD06369648
  • Cas Number : 851175-22-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11583

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11583
Availability details for
Quantity
  • Availability
  • Add to Cart