ENA048303561 Enamine

1-[3,5-bis(furan-2-yl)-4,5-dihydro-1H-pyrazol-1-yl]-2-chloroethan-1-one

  • MDL number MFCD06371776
  • Cas Number : 796084-57-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11590

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11590
Availability details for
Quantity
  • Availability
  • Add to Cart