ENA048303570 Enamine

2-{[5-methyl-4-(4-methylphenyl)-4H-1,2,4-triazol-3-yl]sulfanyl}acetic acid

  • MDL number MFCD06370786
  • Cas Number : 796067-59-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-11603

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11603
Availability details for
Quantity
  • Availability
  • Add to Cart