ENA048303573 Enamine

7-chloro-3-(3-ethoxypropyl)-2-mercaptoquinazolin-4(3H)-one

  • MDL number MFCD06370789
  • Cas Number : 796067-61-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11607

Safety Information

msds https://www.enaminestore.com/catalog/EN300-11607
Availability details for
Quantity
  • Availability
  • Add to Cart