ENA048303574 Enamine

1-[(5-chlorothiophen-2-yl)methyl]piperazine

  • MDL number MFCD06370793
  • Cas Number : 55513-18-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-11608

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11608
Availability details for
Quantity
  • Availability
  • Add to Cart