ENA048303583 Enamine

4-chloro-5,6-dimethyl-2-(thiophen-2-yl)thieno[2,3-d]pyrimidine

  • MDL number MFCD06375946
  • Cas Number : 851208-04-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 130-137
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-11624

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11624
Availability details for
Quantity
  • Availability
  • Add to Cart