ENA048303586 Enamine

5-(4-bromophenyl)-1-(2-methylpropyl)-1H-imidazole-2-thiol

  • MDL number MFCD06371767
  • Cas Number : 796084-50-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11632

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11632
Availability details for
Quantity
  • Availability
  • Add to Cart