ENA048303599 Enamine

2-(chloromethyl)-5-(2,5-dimethylfuran-3-yl)-1,3,4-oxadiazole

  • MDL number MFCD06372760
  • Cas Number : 796106-49-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 62-63
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11649

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11649
Availability details for
Quantity
  • Availability
  • Add to Cart