ENA048303601 Enamine

2-(chloromethyl)-5-(4,5,6,7-tetrahydro-1-benzothiophen-2-yl)-1,3,4-oxadiazole

  • MDL number MFCD06372768
  • Cas Number : 796106-53-1
  • Molecular Weight : 254.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 175-176
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-11651

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11651
Availability details for
Quantity
  • Availability
  • Add to Cart