ENA048303611 Enamine

5-amino-2-methoxy-N,N-dimethylbenzene-1-sulfonamide

  • MDL number MFCD00025827
  • Cas Number : 851175-92-3
  • Molecular Weight : 230.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 132-133
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11668

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11668
Availability details for
Quantity
  • Availability
  • Add to Cart