ENA048303628 Enamine

N-(2-chloroacetyl)thiophene-2-carboxamide

  • MDL number MFCD06373479
  • Cas Number : 851176-02-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11701

Safety Information

msds https://www.enaminestore.com/catalog/EN300-11701
Availability details for
Quantity
  • Availability
  • Add to Cart