ENA048303642 Enamine

2-[(5-chlorothiophen-2-yl)methanesulfonyl]acetic acid

  • MDL number MFCD06373469
  • Cas Number : 851175-94-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11727

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11727
Availability details for
Quantity
  • Availability
  • Add to Cart