ENA048303643 Enamine

4-chloro-3-[(prop-2-en-1-yl)sulfamoyl]benzoic acid

  • MDL number MFCD06363742
  • Cas Number : 59210-69-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-11317

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11317
Availability details for
Quantity
  • Availability
  • Add to Cart