ENA048303657 Enamine

1-(prop-2-en-1-yl)-1H-1,3-benzodiazole-2-thiol

  • MDL number MFCD06655204
  • Cas Number : 87216-53-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10964

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10964
Availability details for
Quantity
  • Availability
  • Add to Cart