ENA048303658 Enamine

2-[(adamantan-1-yl)formamido]propanoic acid

  • Cas Number : 237400-97-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11332

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11332
Availability details for
Quantity
  • Availability
  • Add to Cart