ENA048303660 Enamine

3-hydroxy-2-(4-methoxybenzenesulfonamido)butanoic acid

  • MDL number MFCD05270895
  • Cas Number : 113793-31-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-09385

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-09385
Availability details for
Quantity
  • Availability
  • Add to Cart