ENA048303678 Enamine

2-(4-methoxybenzenesulfonamido)-4-methylpentanoic acid

  • MDL number MFCD05148327
  • Cas Number : 202751-32-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11352

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11352
Availability details for
Quantity
  • Availability
  • Add to Cart