ENA048303680 Enamine

3-(1H-indol-3-yl)-2-{[(3-methylphenyl)sulfonyl]amino}propanoic acid

  • MDL number MFCD06364932

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11354

Safety Information

msds https://www.enaminestore.com/catalog/EN300-11354
Availability details for
Quantity
  • Availability
  • Add to Cart