ENA048303691 Enamine

4-(2,3-dihydro-1H-inden-5-yl)-4-oxobutanoic acid

  • MDL number MFCD03787570
  • Cas Number : 75382-32-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 96-97
Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11365

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11365
Availability details for
Quantity
  • Availability
  • Add to Cart