ENA048303693 Enamine

1,3-diphenyl-4,5-dihydro-1H-pyrazol-5-one

  • MDL number MFCD00117375
  • Cas Number : 4845-49-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-05386

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-05386
Availability details for
Quantity
  • Availability
  • Add to Cart