ENA048303695 Enamine

6-chloro-2-hydrazinyl-4-methylquinoline

  • MDL number MFCD06254046
  • Cas Number : 21703-54-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11369

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11369
Availability details for
Quantity
  • Availability
  • Add to Cart