ENA048303698 Enamine

2-[(4-chloro-3-nitrophenyl)formamido]propanoic acid

  • MDL number MFCD02323130
  • Molecular Weight : 272.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-09916

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-09916
Availability details for
Quantity
  • Availability
  • Add to Cart