ENA048303702 Enamine

6-bromo-3-(2-cyclohex-1-en-1-ylethyl)-2-mercaptoquinazolin-4(3H)-one

  • MDL number MFCD06358632
  • Cas Number : 403727-60-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11118

Safety Information

msds https://www.enaminestore.com/catalog/EN300-11118
Availability details for
Quantity
  • Availability
  • Add to Cart