ENA048303705 Enamine

methyl 4-oxo-3-[(oxolan-2-yl)methyl]-2-sulfanyl-3,4-dihydroquinazoline-7-carboxylate

  • MDL number MFCD06358645
  • Cas Number : 422526-41-8
  • Molecular Weight : 320.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-11120

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11120
Availability details for
Quantity
  • Availability
  • Add to Cart