ENA048303742 Enamine

2-chloro-N-{5-[(3-methylphenyl)methyl]-1,3-thiazol-2-yl}acetamide

  • MDL number MFCD02279389
  • Cas Number : 450360-89-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 130-131
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11139

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11139
Availability details for
Quantity
  • Availability
  • Add to Cart