ENA048303749 Enamine

4-oxo-4-(5,6,7,8-tetrahydronaphthalen-2-yl)butanoic acid

  • MDL number MFCD06655207
  • Cas Number : 785-17-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 107-108
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10990

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10990
Availability details for
Quantity
  • Availability
  • Add to Cart