ENA048303750 Enamine

1-(1-methylpiperidin-4-yl)-1H-pyrazol-5-amine

  • MDL number MFCD03856631
  • Cas Number : 3524-30-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 98...100
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11425

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-11425
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart