ENA048303753 Enamine

2-chloro-1-(2-chloro-10H-phenothiazin-10-yl)ethan-1-one

  • MDL number MFCD02087295
  • Cas Number : 16189-69-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11145

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11145
Availability details for
Quantity
  • Availability
  • Add to Cart