ENA048303759 Enamine

3-(4-methoxybenzoyl)-4,5,6,7-tetrahydro-1-benzothiophen-2-amine

  • MDL number MFCD00427061
  • Cas Number : 40312-30-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11434

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11434
Availability details for
Quantity
  • Availability
  • Add to Cart