ENA048303760 Enamine

2-[2-(3-chlorophenyl)-1,3-thiazol-4-yl]acetic acid

  • MDL number MFCD06366743
  • Cas Number : 17969-26-5
  • Molecular Weight : 253.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp 99-100
Iupac Name [2-(3-chlorophenyl)-1,3-thiazol-4-yl]acetic acid
Inchi Code 1S/C11H8ClNO2S/c12-8-3-1-2-7(4-8)11-13-9(6-16-11)5-10(14)15/h1-4,6H,5H2,(H,14,15)
Inchi Key CCUFHQQWZTVOCR-UHFFFAOYSA-N

Description

Melting Point 99-100
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-11435

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-11435
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart