ENA048303764 Enamine

2-[2-(3-fluorophenyl)-1,3-thiazol-4-yl]acetic acid

  • MDL number MFCD06366742
  • Cas Number : 794554-74-8
  • Molecular Weight : 237.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11439

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11439
Availability details for
Quantity
  • Availability
  • Add to Cart