ENA048303768 Enamine

2-bromo-5-{[(furan-2-yl)methyl]sulfamoyl}benzoic acid

  • MDL number MFCD06359552
  • Cas Number : 790719-73-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-11150

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11150
Availability details for
Quantity
  • Availability
  • Add to Cart