ENA048303770 Enamine

2-chloro-1-[2,5-dimethyl-1-(3-nitrophenyl)-1H-pyrrol-3-yl]ethan-1-one

  • MDL number MFCD06253999
  • Cas Number : 610274-32-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-11151

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11151
Availability details for
Quantity
  • Availability
  • Add to Cart