ENA048303771 Enamine

2-chloro-1-{1-[2-(3,4-dimethoxyphenyl)ethyl]-2,5-dimethyl-1H-pyrrol-3-yl}ethanone

  • MDL number MFCD06253998
  • Cas Number : 610274-31-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11152

Safety Information

msds https://www.enaminestore.com/catalog/EN300-11152
Availability details for
Quantity
  • Availability
  • Add to Cart