ENA048303776 Enamine

[3-(carboxymethyl)-2-oxo-2,3-dihydro-1H-benzimidazol-1-yl]acetic acid

  • MDL number MFCD00477924
  • Cas Number : 1848-99-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11451

Safety Information

msds https://www.enaminestore.com/catalog/EN300-11451
Availability details for
Quantity
  • Availability
  • Add to Cart