ENA048303781 Enamine

2-(4-chlorobenzamido)benzoic acid

  • MDL number MFCD00276714
  • Cas Number : 34425-87-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11456

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11456
Availability details for
Quantity
  • Availability
  • Add to Cart