ENA048303792 Enamine

2-phenyl-2H-1,2,3-triazole-4-carbaldehyde

  • MDL number MFCD01104763
  • Cas Number : 3213-80-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 66...68
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11467

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-11467
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart