ENA048303794 Enamine

5-(4-methylphenyl)thieno[2,3-d]pyrimidine-4-thiol

  • MDL number MFCD04971692
  • Cas Number : 307512-34-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10997

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10997
Availability details for
Quantity
  • Availability
  • Add to Cart