ENA048303796 Enamine

2-phenyl-2H-1,2,3-triazole-4-carboxylic acid

  • MDL number MFCD01104530
  • Cas Number : 13306-99-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 184-185
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11471

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11471
Availability details for
Quantity
  • Availability
  • Add to Cart