ENA048303800 Enamine

(2S)-2-{[(4-methoxyphenyl)sulfonyl]amino}propanoic acid

  • MDL number MFCD00664347
  • Cas Number : 85622-69-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08320

Safety Information

msds https://www.enaminestore.com/catalog/EN300-08320
Availability details for
Quantity
  • Availability
  • Add to Cart